siRNA Details
Gene Symbol: REG3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008387.2 | NP_001008388.1 | 5, 6 | 658 | ||
NM_001270040.1 | NP_001256969.1 | 4, 5 | 520 | |||
NM_198448.3 | NP_940850.1 | 5, 6 | 558 | |||
XM_005264135.2 | XP_005264192.1 | 5, 6 | 720 | |||
GenBank mRNA | AB161037.1 | - | 5, 6 | 543 | ||
AB161038.1 | - | 5, 6 | 643 | |||
AK292595.1 | - | 5, 6 | 542 | |||
AY359047.1 | - | 5, 6 | 544 | |||
AY428734.1 | - | 5, 6 | 517 | |||
BC103852.2 | - | 4, 5 | 479 | 40002928 | ||
BC103853.1 | - | 5, 6 | 617 | 40002929 | ||
BC103854.1 | - | 5, 6 | 517 | 40002931 | ||
CR541938.1 | - | 4, 5 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
