siRNA Details
Gene Symbol: REG3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008387.2 | NP_001008388.1 | 6 | 858 | ||
NM_001270040.1 | NP_001256969.1 | 5 | 720 | |||
NM_198448.3 | NP_940850.1 | 6 | 758 | |||
XM_005264135.2 | XP_005264192.1 | 6 | 920 | |||
GenBank mRNA | AB161037.1 | - | 6 | 743 | ||
AB161038.1 | - | 6 | 843 | |||
AK292595.1 | - | 6 | 742 | |||
AY359047.1 | - | 6 | 744 | |||
AY428734.1 | - | 6 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
