siRNA Details
Gene Symbol: PRSS56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195129.1 | NP_001182058.1 | 5 | 645 | ||
GenBank mRNA | BI464671.1 | - | 2 | 199 | ||
CN280933.1 | - | 5 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
