siRNA Details
Gene Symbol: RBMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003793.2 | NP_001003793.1 | 9, 10 | 1259 | ||
NM_001177711.1 | NP_001171182.1 | 9, 10 | 1259 | |||
NM_001177712.1 | NP_001171183.1 | 10, 11 | 1298 | |||
NM_014483.3 | NP_055298.2 | 9, 10 | 1259 | |||
XM_005265060.2 | XP_005265117.1 | 9, 10 | 1708 | |||
XM_005265063.1 | XP_005265120.1 | 6, 7 | 916 | |||
XM_005265065.4 | XP_005265122.1 | 6, 7 | 609 | |||
XM_017006178.1 | XP_016861667.1 | 9, 10 | 1708 | |||
XM_017006179.1 | XP_016861668.1 | 9, 10 | 1708 | |||
GenBank mRNA | AF023259.1 | - | 9, 10 | 1036 | ||
AK292789.1 | - | 9, 10 | 935 | |||
AY236871.1 | - | 9, 10 | 1119 | |||
BC117315.1 | - | 10, 11 | 995 | 40125866 | ||
BC143521.1 | - | 10, 11 | 995 | 9052030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
