siRNA Details
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 4 | 502 | ||
NM_001007073.1 | NP_001007074.1 | 4 | 601 | |||
NM_001007074.1 | NP_001007075.1 | 3 | 621 | |||
GenBank mRNA | AK124029.1 | - | 1 | 2316 | ||
BC011514.1 | - | 1 | 428 | 2900089 | ||
BC070209.1 | - | 1 | 434 | 6738371 | ||
BQ068496.1 | - | 539 | ||||
CF124158.1 | - | 4 | 539 | |||
X03342.1 | - | 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
