siRNA Details
Gene Symbol: TAGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008272.1 | NP_001008273.1 | 5 | 1172 | ||
NM_001008273.1 | NP_001008274.1 | 4 | 1057 | |||
NM_013259.2 | NP_037391.2 | 5 | 1346 | |||
GenBank mRNA | AF087855.1 | - | 5 | 1055 | ||
AF112201.1 | - | 4 | 844 | |||
AF303058.1 | - | 5 | 1211 | |||
AK315289.1 | - | 5 | 1030 | |||
BC015329.1 | - | 5 | 928 | 4157360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
