siRNA Details
Gene Symbol: TAGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008272.1 | NP_001008273.1 | 3 | 589 | ||
NM_001008273.1 | NP_001008274.1 | 2 | 474 | |||
NM_013259.2 | NP_037391.2 | 3 | 763 | |||
GenBank mRNA | AF087855.1 | - | 3 | 475 | ||
AF112201.1 | - | 2 | 260 | |||
AF303058.1 | - | 3 | 628 | |||
AK315289.1 | - | 3 | 447 | |||
BC015329.1 | - | 3 | 345 | 4157360 | ||
BG912462.1 | - | 2 | 479 | |||
BI752099.1 | - | 3 | 664 | |||
BI822856.1 | - | 2 | 410 | |||
BT007408.1 | - | 2 | 211 | |||
CK003985.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
