siRNA Details
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 18 | 5245 | ||
XM_005263073.3 | XP_005263130.1 | 17 | 5119 | |||
XM_005263075.2 | XP_005263132.1 | 17 | 5134 | |||
XM_005263076.2 | XP_005263133.1 | 17 | 5131 | |||
XM_005263077.3 | XP_005263134.1 | 17 | 5187 | |||
XM_006714240.1 | XP_006714303.1 | 16 | 4320 | |||
XM_011532037.2 | XP_011530339.1 | 18 | 5297 | |||
XM_011532038.2 | XP_011530340.1 | 17 | 4574 | |||
XM_017008312.1 | XP_016863801.1 | 19 | 5411 | |||
XM_017008313.1 | XP_016863802.1 | 18 | 5096 | |||
XM_017008314.1 | XP_016863803.1 | 18 | 5095 | |||
XM_017008315.1 | XP_016863804.1 | 18 | 5165 | |||
XM_017008316.1 | XP_016863805.1 | 16 | 4983 | |||
XM_017008317.1 | XP_016863806.1 | 16 | 5011 | |||
XM_017008318.1 | XP_016863807.1 | 17 | 5133 | |||
GenBank mRNA | AK022799.1 | - | 2 | 1911 | ||
BC017382.1 | - | 1 | 1574 | |||
BX648339.1 | - | 16 | 4333 | |||
CR749490.1 | - | 19 | 5360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
