siRNA Details
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003381.3 | NP_003372.1 | 4 | 493 | ||
NM_194435.2 | NP_919416.1 | 4 | 493 | |||
XM_005267135.3 | XP_005267192.1 | 4 | 445 | |||
XM_006715562.3 | XP_006715625.1 | 4 | 3396 | |||
GenBank mRNA | AK291959.1 | - | 4 | 445 | ||
AK313950.1 | - | 4 | 493 | |||
BC009794.1 | - | 4 | 498 | 4294373 | ||
DA914915.1 | - | 4 | 493 | |||
M36634.1 | - | 4 | 386 | |||
M54930.1 | - | 2 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
