siRNA Details
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 6 | 979 | ||
GenBank mRNA | AF035277.1 | - | 6 | 827 | ||
AK090513.1 | - | 4 | 1810 | |||
AK315122.1 | - | 6 | 849 | |||
BC010550.1 | - | 6 | 780 | 4153601 | ||
BX647744.1 | - | 2 | 965 | |||
CR456803.1 | - | 6 | 715 | |||
CR541850.1 | - | 6 | 715 | |||
KU178337.1 | - | 6 | 715 | |||
S79669.1 | - | 6 | 867 | |||
U17280.1 | - | 6 | 841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
