siRNA Details
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 5 | 746 | ||
XM_006716392.2 | XP_006716455.1 | 5 | 929 | |||
GenBank mRNA | AF035277.1 | - | 5 | 594 | ||
AK090513.1 | - | 4 | 930 | |||
AK315122.1 | - | 5 | 616 | |||
BC010550.1 | - | 5 | 547 | 4153601 | ||
CR456803.1 | - | 5 | 482 | |||
CR541850.1 | - | 5 | 482 | |||
KU178337.1 | - | 5 | 482 | |||
S79669.1 | - | 5 | 634 | |||
U17280.1 | - | 5 | 608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
