siRNA Details
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 7 | 1695 | ||
XM_006716392.2 | XP_006716455.1 | 6 | 1784 | |||
GenBank mRNA | AF035277.1 | - | 7 | 1543 | ||
AK090513.1 | - | 4 | 3359 | |||
AV706872.1 | - | 1 | 193 | |||
BC010550.1 | - | 7 | 1496 | 4153601 | ||
BX647744.1 | - | 2 | 2516 | |||
S79669.1 | - | 7 | 1584 | |||
U17280.1 | - | 7 | 1556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
