siRNA Details
Gene Symbol: TRIM55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033058.2 | NP_149047.2 | 1 | 276 | ||
NM_184085.1 | NP_908973.1 | 1 | 276 | |||
NM_184086.1 | NP_908974.1 | 1 | 276 | |||
NM_184087.1 | NP_908975.1 | 1 | 276 | |||
XM_005251316.4 | XP_005251373.1 | 3 | 1301 | |||
XM_011517613.2 | XP_011515915.1 | 3 | 1301 | |||
XM_011517614.2 | XP_011515916.1 | 3 | 1224 | |||
XM_011517615.2 | XP_011515917.1 | 3 | 1301 | |||
XM_011517616.2 | XP_011515918.1 | 3 | 1301 | |||
GenBank mRNA | AJ243488.1 | - | 1 | 276 | ||
AJ243489.1 | - | 1 | 276 | |||
AJ277493.1 | - | 1 | 276 | |||
AJ291712.1 | - | 1 | 182 | |||
AJ431704.1 | - | 1 | 276 | |||
AK091310.1 | - | 1 | 426 | |||
AK091728.1 | - | 1 | 96 | |||
BC007750.2 | - | 1 | 182 | 4110783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
