siRNA Details
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024013.2 | NP_076918.1 | 1 | 632 | ||
GenBank mRNA | BC074928.2 | - | 1 | 608 | 30915487 | |
BC074929.2 | - | 1 | 608 | 30915263 | ||
BC112300.1 | - | 1 | 616 | 8327768 | ||
BC112302.1 | - | 1 | 616 | 8327770 | ||
V00537.1 | - | 1 | 634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
