siRNA Details
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 16 | 1368 | ||
NM_001145356.1 | NP_001138828.1 | 14 | 1213 | |||
NM_018491.3 | NP_060961.3 | 15 | 1270 | |||
XM_005251513.4 | XP_005251570.1 | 14 | 1218 | |||
XM_005251514.1 | XP_005251571.1 | 14 | 1183 | |||
XM_005251515.1 | XP_005251572.1 | 13 | 1126 | |||
XM_011517961.2 | XP_011516263.1 | 15 | 1305 | |||
XM_011517962.2 | XP_011516264.1 | 14 | 1140 | |||
XM_011517963.1 | XP_011516265.1 | 12 | 808 | |||
GenBank mRNA | AF257330.2 | - | 15 | 1228 | ||
AK290648.1 | - | 16 | 1340 | |||
AK315855.1 | - | 15 | 1194 | |||
AY343911.1 | - | 15 | 1227 | |||
BC005996.1 | - | 15 | 1270 | 4069589 | ||
BC009573.1 | - | 16 | 1321 | 4133536 | ||
BC013432.1 | - | 15 | 1241 | 4133722 | ||
BC132759.1 | - | 15 | 1204 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225251.1 | - | 16 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
