siRNA Details
Gene Symbol: OR1K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080859.1 | NP_543135.1 | 1 | 339 | ||
GenBank mRNA | BC137033.1 | - | 1 | 371 | 9021029 | |
BC137034.1 | - | 1 | 371 | 9021030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
