siRNA Details
Gene Symbol: OR5C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001923.1 | NP_001001923.1 | 1 | 589 | ||
GenBank mRNA | BC136700.1 | - | 1 | 651 | 9020690 | |
BC136701.1 | - | 1 | 651 | 9020691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
