siRNA Details
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202403.1 | NP_001189332.1 | 5 | 4484 | ||
NM_017873.3 | NP_060343.1 | 6 | 4571 | |||
NM_177999.2 | NP_821066.1 | 5 | 4462 | |||
GenBank mRNA | AK093539.1 | - | 1 | 2119 | ||
AL390135.1 | - | 1 | 2844 | |||
BX648923.1 | - | 5 | 5093 | |||
CR627462.1 | - | 1 | 2844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
