siRNA Details
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202403.1 | NP_001189332.1 | 5 | 1123 | ||
NM_017873.3 | NP_060343.1 | 6 | 1210 | |||
NM_177999.2 | NP_821066.1 | 5 | 1101 | |||
GenBank mRNA | AK000555.1 | - | 6 | 1207 | ||
AK023004.1 | - | 6 | 1156 | |||
AK225618.1 | - | 5 | 1098 | |||
AK292807.1 | - | 6 | 1207 | |||
AK299193.1 | - | 5 | 1005 | |||
BC001719.1 | - | 5 | 1056 | |||
BC065913.1 | - | 6 | 1113 | 5211288 | ||
BX648923.1 | - | 5 | 1731 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
