siRNA Details
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004470.1 | NP_001004470.1 | 2, 3 | 202 | ||
GenBank mRNA | AB062101.1 | - | 2, 3 | 202 | ||
AJ621583.1 | - | 2, 3 | 202 | |||
BC137102.1 | - | 2, 3 | 334 | 9021099 | ||
BC137103.1 | - | 2, 3 | 334 | 9021100 | ||
BC144472.1 | - | 2, 3 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
