siRNA Details
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001971.2 | NP_001001971.1 | 12 | 1607 | ||
NM_001143773.1 | NP_001137245.1 | 15 | 2072 | |||
NM_001166698.1 | NP_001160170.1 | 15 | 1983 | |||
NM_198215.3 | NP_937858.2 | 14 | 1901 | |||
XM_005269618.4 | XP_005269675.2 | 14 | 1898 | |||
XM_005269619.4 | XP_005269676.2 | 12 | 1604 | |||
XM_006717702.3 | XP_006717765.1 | 16 | 1902 | |||
XM_006717703.3 | XP_006717766.1 | 16 | 2084 | |||
XM_011539490.2 | XP_011537792.1 | 15 | 1957 | |||
XM_011539491.2 | XP_011537793.1 | 16 | 2003 | |||
XM_017015885.1 | XP_016871374.1 | 15 | 1815 | |||
XM_017015886.1 | XP_016871375.1 | 15 | 2069 | |||
XM_017015887.1 | XP_016871376.1 | 15 | 1819 | |||
XM_017015888.1 | XP_016871377.1 | 16 | 2081 | |||
XM_017015889.1 | XP_016871378.1 | 16 | 1899 | |||
XM_017015890.1 | XP_016871379.1 | 16 | 2138 | |||
XM_017015891.1 | XP_016871380.1 | 13 | 1778 | |||
XM_017015892.1 | XP_016871381.1 | 13 | 1775 | |||
XM_017015893.1 | XP_016871382.1 | 15 | 1879 | |||
GenBank mRNA | AK289796.1 | - | 14 | 1827 | ||
AK299924.1 | - | 15 | 2072 | |||
AK316542.1 | - | 15 | 1979 | |||
BC036453.1 | - | 14 | 1859 | 5270033 | ||
BC064431.1 | - | 12 | 1607 | 5764753 |
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW299583.1 | - | 1 | 173 | ||
BM676577.1 | - | 1 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
