siRNA Details
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001971.2 | NP_001001971.1 | 1, 2 | 195 | ||
NM_001143773.1 | NP_001137245.1 | 2, 3 | 366 | |||
NM_198215.3 | NP_937858.2 | 1, 2 | 195 | |||
XM_005269618.4 | XP_005269675.2 | 1, 2 | 195 | |||
XM_005269619.4 | XP_005269676.2 | 1, 2 | 195 | |||
XM_011539490.2 | XP_011537792.1 | 1, 2 | 162 | |||
XM_017015885.1 | XP_016871374.1 | 2, 3 | 109 | |||
XM_017015886.1 | XP_016871375.1 | 2, 3 | 366 | |||
XM_017015887.1 | XP_016871376.1 | 2, 3 | 116 | |||
XM_017015890.1 | XP_016871379.1 | 2, 3 | 346 | |||
XM_017015891.1 | XP_016871380.1 | 2, 3 | 366 | |||
XM_017015892.1 | XP_016871381.1 | 2, 3 | 366 | |||
GenBank mRNA | AK289796.1 | - | 1, 2 | 121 | ||
AK294805.1 | - | 1, 2 | 110 | |||
AK299924.1 | - | 2, 3 | 366 | |||
BC036453.1 | - | 1, 2 | 153 | 5270033 | ||
BC064431.1 | - | 1, 2 | 195 | 5764753 | ||
DA188937.1 | - | 1, 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
