siRNA Details
Gene Symbol: OR5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003697.1 | NP_003688.1 | 1 | 558 | ||
GenBank mRNA | BC101239.2 | - | 1 | 560 | 40023020 | |
BC101241.1 | - | 1 | 558 | |||
BC101242.3 | - | 1 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
