siRNA Details
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 11 | 1462 | ||
NM_004879.4 | NP_004870.3 | 11 | 1504 | |||
NR_110769.1 | 10 | 1358 | ||||
NR_110770.1 | 12 | 1629 | ||||
XM_011543069.1 | XP_011541371.1 | 12 | 1520 | |||
XM_011543070.1 | XP_011541372.1 | 10 | 1563 | |||
XM_017018582.1 | XP_016874071.1 | 11 | 1408 | |||
GenBank mRNA | AB209820.1 | - | 11 | 1461 | ||
AF131850.1 | - | 1 | 1270 | |||
AK223029.1 | - | 11 | 1433 | |||
AK296620.1 | - | 11 | 1391 | |||
AK301556.1 | - | 10 | 1287 | |||
AK315841.1 | - | 11 | 1420 | |||
BC002390.2 | - | 11 | 1370 | 2819783 | ||
BC029333.1 | - | 3 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
