siRNA Details
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 11 | 1722 | ||
NM_004879.4 | NP_004870.3 | 11 | 1764 | |||
NR_110769.1 | 10 | 1618 | ||||
NR_110770.1 | 12 | 1889 | ||||
XM_011543069.1 | XP_011541371.1 | 12 | 1780 | |||
XM_011543070.1 | XP_011541372.1 | 10 | 1823 | |||
XM_017018582.1 | XP_016874071.1 | 11 | 1668 | |||
GenBank mRNA | AB209820.1 | - | 11 | 1721 | ||
AF010313.2 | - | 11 | 1636 | |||
AF131850.1 | - | 1 | 1530 | |||
AK223029.1 | - | 11 | 1693 | |||
BC002390.2 | - | 11 | 1630 | 2819783 | ||
BC029333.1 | - | 3 | 771 | |||
BM467354.1 | - | 60 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
