siRNA Details
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 5 | 517 | ||
NM_004879.4 | NP_004870.3 | 5 | 559 | |||
NR_110769.1 | 4 | 413 | ||||
NR_110770.1 | 5 | 513 | ||||
XM_011543069.1 | XP_011541371.1 | 6 | 575 | |||
XM_011543070.1 | XP_011541372.1 | 5 | 730 | |||
XM_017018582.1 | XP_016874071.1 | 6 | 575 | |||
GenBank mRNA | AB209820.1 | - | 5 | 516 | ||
AF010313.2 | - | 5 | 431 | |||
AK223029.1 | - | 5 | 488 | |||
AK291951.1 | - | 5 | 468 | |||
AK296620.1 | - | 5 | 446 | |||
AK301556.1 | - | 4 | 342 | |||
AK315841.1 | - | 5 | 475 | |||
AK316539.1 | - | 5 | 423 | |||
BC002390.2 | - | 5 | 425 | 2819783 | ||
BM450796.1 | - | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
