siRNA Details
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006597.5 | NP_006588.1 | 4 | 827 | ||
NM_153201.3 | NP_694881.1 | 4 | 827 | |||
XM_011542798.1 | XP_011541100.1 | 4 | 575 | |||
GenBank mRNA | AB034951.1 | - | 4 | 591 | ||
AB073886.1 | - | 3 | 505 | |||
AF352832.1 | - | 4 | 614 | |||
AK096100.1 | - | 4 | 553 | |||
AK129885.1 | - | 4 | 629 | |||
AK222628.1 | - | 4 | 625 | |||
AK292067.1 | - | 1 | 257 | |||
AK300404.1 | - | 4 | 701 | |||
AK303935.1 | - | 3 | 398 | |||
AK310467.1 | - | 4 | 699 | |||
BC007276.1 | - | 3 | 375 | |||
BC016179.1 | - | 4 | 608 | 3920744 | ||
BC016660.1 | - | 4 | 608 | 3923981 | ||
BC019816.2 | - | 4 | 608 | 2899894 | ||
EU668349.1 | - | 4 | 608 | |||
FJ224294.1 | - | 4 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
