siRNA Details
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006597.5 | NP_006588.1 | 5 | 1089 | ||
NM_153201.3 | NP_694881.1 | 5 | 1089 | |||
XM_011542798.1 | XP_011541100.1 | 5 | 837 | |||
GenBank mRNA | AB034951.1 | - | 5 | 853 | ||
AB073886.1 | - | 4 | 767 | |||
AF352832.1 | - | 5 | 876 | |||
AK096100.1 | - | 5 | 815 | |||
AK129885.1 | - | 5 | 891 | |||
AK222628.1 | - | 5 | 887 | |||
AK222785.1 | - | 5 | 887 | |||
AK292067.1 | - | 2 | 519 | |||
AK303935.1 | - | 4 | 660 | |||
AK310467.1 | - | 5 | 961 | |||
BC007276.1 | - | 4 | 637 | |||
BC016179.1 | - | 5 | 870 | 3920744 | ||
BC016660.1 | - | 5 | 870 | 3923981 | ||
BC019816.2 | - | 5 | 870 | 2899894 | ||
EU668349.1 | - | 5 | 870 | |||
FJ224294.1 | - | 5 | 890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
