siRNA Details
Gene Symbol: FEZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005103.4 | NP_005094.1 | 9 | 1379 | ||
XM_005271734.2 | XP_005271791.1 | 9 | 1424 | |||
XM_005271735.2 | XP_005271792.1 | 9 | 1476 | |||
GenBank mRNA | BC009545.1 | - | 9 | 1296 | 3897353 | |
CR456817.1 | - | 8 | 1144 | |||
CR541954.1 | - | 8 | 1144 | |||
U60060.1 | - | 9 | 1243 | |||
U69139.1 | - | 8 | 1169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
