siRNA Details
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004608.3 | NP_004599.2 | 5, 6 | 897 | ||
XM_005255523.2 | XP_005255580.1 | 6, 7 | 1182 | |||
XM_011545926.2 | XP_011544228.1 | 6, 7 | 1345 | |||
XM_017023614.1 | XP_016879103.1 | 6, 7 | 922 | |||
GenBank mRNA | AJ007989.1 | - | 5, 6 | 861 | ||
BC026031.1 | - | 5, 6 | 897 | 5460190 | ||
KJ534981.1 | - | 5, 6 | 933 | |||
KJ535035.1 | - | 4, 5 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
