siRNA Details
Gene Symbol: CNTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243270.1 | NP_001230199.1 | 5 | 814 | ||
NM_001243271.1 | NP_001230200.1 | 6 | 953 | |||
NM_014361.3 | NP_055176.1 | 6 | 953 | |||
NM_175566.2 | NP_780775.1 | 3 | 297 | |||
XM_011542871.1 | XP_011541173.1 | 5 | 731 | |||
XM_011542873.1 | XP_011541175.1 | 6 | 953 | |||
XM_017017926.1 | XP_016873415.1 | 7 | 1121 | |||
XM_017017927.1 | XP_016873416.1 | 7 | 1121 | |||
XM_017017928.1 | XP_016873417.1 | 5 | 886 | |||
XM_017017929.1 | XP_016873418.1 | 5 | 731 | |||
GenBank mRNA | AB013802.2 | - | 4 | 528 | ||
AB013803.2 | - | 3 | 306 | |||
AK308788.1 | - | 6 | 953 | |||
BC039255.1 | - | 6 | 788 | 5273769 | ||
BC130619.1 | - | 5 | 600 | 40146650 | ||
BC144185.1 | - | 5 | 600 | 9052705 | ||
BC144186.1 | - | 6 | 739 | 9052706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
