siRNA Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039661.1 | NP_001034750.1 | 6 | 1414 | ||
NM_001318777.1 | NP_001305706.1 | 5 | 1261 | |||
XM_011542576.2 | XP_011540878.1 | 4 | 1159 | |||
XM_017017161.1 | XP_016872650.1 | 3 | 2782 | |||
XM_017017162.1 | XP_016872651.1 | 6 | 1451 | |||
GenBank mRNA | AF410783.1 | - | 7 | 1218 | ||
AK124298.1 | - | 5 | 1261 | |||
AY576785.1 | - | 3 | 1017 | |||
AY576786.1 | - | 3 | 1017 | |||
BC032474.1 | - | 6 | 1383 | 5216171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
