siRNA Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039661.1 | NP_001034750.1 | 5, 6 | 1077 | ||
NM_001318777.1 | NP_001305706.1 | 4, 5 | 924 | |||
XM_011542576.2 | XP_011540878.1 | 3, 4 | 822 | |||
XM_017017161.1 | XP_016872650.1 | 2, 3 | 2445 | |||
XM_017017162.1 | XP_016872651.1 | 5, 6 | 1114 | |||
GenBank mRNA | AB446477.1 | - | 2, 3 | 641 | ||
AF410783.1 | - | 5, 6 | 1070 | |||
AK124298.1 | - | 4, 5 | 924 | |||
AY576785.1 | - | 2, 3 | 680 | |||
AY576786.1 | - | 2, 3 | 680 | |||
BC032474.1 | - | 5, 6 | 1046 | 5216171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
