siRNA Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258317.3 | XP_005258374.1 | 2 | 436 | ||
XM_017025876.1 | XP_016881365.1 | 2 | 357 | |||
GenBank mRNA | AB037749.2 | - | 2 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
