siRNA Details
Gene Symbol: CTAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247988.1 | NP_001234917.1 | 23 | 3302 | ||
NM_001247989.1 | NP_001234918.1 | 24 | 3615 | |||
NM_001247990.1 | NP_001234919.1 | 24 | 3505 | |||
NM_005930.3 | NP_005921.2 | 24 | 3600 | |||
NM_203354.2 | NP_976229.1 | 24 | 3351 | |||
NM_203355.2 | NP_976230.1 | 23 | 3471 | |||
NM_203356.2 | NP_976231.1 | 24 | 3431 | |||
GenBank mRNA | U94780.1 | - | 24 | 3580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
