siRNA Details
Gene Symbol: WDR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170710.4 | NP_733828.2 | 4 | 562 | ||
NM_181265.3 | NP_851782.3 | 3 | 439 | |||
XM_011531582.2 | XP_011529884.1 | 3 | 427 | |||
XM_017007686.1 | XP_016863175.1 | 3 | 415 | |||
XM_017007687.1 | XP_016863176.1 | 3 | 428 | |||
XM_017007688.1 | XP_016863177.1 | 4 | 417 | |||
XM_017007689.1 | XP_016863178.1 | 3 | 414 | |||
XM_017007690.1 | XP_016863179.1 | 3 | 428 | |||
XM_017007691.1 | XP_016863180.1 | 3 | 414 | |||
XM_017007692.1 | XP_016863181.1 | 3 | 428 | |||
XM_017007693.1 | XP_016863182.1 | 3 | 414 | |||
XM_017007694.1 | XP_016863183.1 | 4 | 398 | |||
XM_017007695.1 | XP_016863184.1 | 3 | 428 | |||
XM_017007696.1 | XP_016863185.1 | 3 | 428 | |||
XM_017007697.1 | XP_016863186.1 | 3 | 416 | |||
GenBank mRNA | AF492460.1 | - | 4 | 415 | ||
AK098748.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
