siRNA Details
Gene Symbol: ATP6V1G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320218.1 | NP_001307147.1 | 4 | 249 | ||
NM_133262.2 | NP_573569.1 | 3 | 231 | |||
NM_133326.1 | NP_579872.1 | 4 | 277 | |||
XM_006711163.3 | XP_006711226.1 | 2 | 236 | |||
XM_011509186.2 | XP_011507488.1 | 3 | 261 | |||
XM_011509187.2 | XP_011507489.1 | 3 | 302 | |||
GenBank mRNA | AY039760.1 | - | 2 | 125 | ||
BC101129.2 | - | 3 | 167 | 40014378 | ||
BC101130.1 | - | 2 | 151 | 40014381 | ||
BC101131.2 | - | 3 | 197 | 40014383 | ||
BX114684.1 | - | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
