siRNA Details
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 28 | 6607 | ||
NM_020779.3 | NP_065830.2 | 27 | 6574 | |||
XM_011533007.2 | XP_011531309.1 | 17 | 5299 | |||
GenBank mRNA | AB037757.2 | - | 28 | 6615 | ||
AK022866.1 | - | 1 | 2202 | |||
BC033951.1 | - | 1 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
