siRNA Details
Gene Symbol: TXNDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160047.1 | NP_001153519.1 | 17, 18 | 2144 | ||
NM_020784.2 | NP_065835.2 | 17, 18 | 2159 | |||
GenBank mRNA | AB037765.2 | - | 17, 18 | 2071 | ||
AK022563.1 | - | 3, 4 | 345 | |||
AK097748.1 | - | 18, 19 | 2048 | |||
BC137081.1 | - | 17, 18 | 2039 | 9021078 | ||
BC137082.1 | - | 17, 18 | 2039 | 9021079 | ||
BC142650.1 | - | 17, 18 | 2082 | 40148019 | ||
BC144469.1 | - | 17, 18 | 2024 | 9052996 | ||
BC150205.1 | - | 17, 18 | 2080 | 40148865 | ||
BC152428.1 | - | 17, 18 | 2071 | 8862542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
