siRNA Details
Gene Symbol: ASTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004319.2 | NP_004310.1 | 23 | 6905 | ||
XM_017001340.1 | XP_016856829.1 | 23 | 6879 | |||
GenBank mRNA | AB006627.1 | - | 23 | 6703 | ||
BC146795.1 | - | 23 | 6703 | 9007266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
