siRNA Details
Gene Symbol: XRCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318012.1 | NP_001304941.1 | 2 | 335 | ||
NM_001318013.1 | NP_001304942.1 | 2 | 288 | |||
NM_003401.4 | NP_003392.1 | 2 | 288 | |||
NM_022406.3 | NP_071801.1 | 2 | 288 | |||
NM_022550.3 | NP_072044.1 | 2 | 335 | |||
XM_011543626.1 | XP_011541928.1 | 2 | 687 | |||
XM_017009827.1 | XP_016865316.1 | 2 | 288 | |||
XM_017009828.1 | XP_016865317.1 | 2 | 288 | |||
XM_017009829.1 | XP_016865318.1 | 2 | 288 | |||
GenBank mRNA | AB017445.1 | - | 2 | 112 | ||
AK290739.1 | - | 2 | 116 | |||
BC005259.1 | - | 2 | 140 | 3838919 | ||
BC016314.1 | - | 2 | 155 | 4108822 | ||
BT007216.1 | - | 1 | 71 | |||
DA067761.1 | - | 2 | 295 | |||
U40622.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
