siRNA Details
Gene Symbol: ANKRD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195144.1 | NP_001182073.1 | 14 | 1557 | ||
XM_005246945.1 | XP_005247002.1 | 14 | 1601 | |||
XM_005246946.1 | XP_005247003.1 | 14 | 1547 | |||
XM_005246947.2 | XP_005247004.1 | 14 | 1601 | |||
XM_005246948.2 | XP_005247005.1 | 14 | 1601 | |||
XM_006712838.3 | XP_006712901.1 | 14 | 1601 | |||
XM_011512148.2 | XP_011510450.1 | 14 | 1601 | |||
XM_017005252.1 | XP_016860741.1 | 14 | 1547 | |||
GenBank mRNA | AK097086.1 | - | 14 | 1366 | ||
AK301209.1 | - | 13 | 1478 | |||
AL133087.1 | - | 8 | 810 | |||
BC016985.2 | - | 14 | 1531 | 4395011 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
