siRNA Details
Gene Symbol: ANKRD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195144.1 | NP_001182073.1 | 10 | 1243 | ||
NM_153697.2 | NP_710181.2 | 10 | 1243 | |||
XM_005246945.1 | XP_005247002.1 | 10 | 1233 | |||
XM_005246946.1 | XP_005247003.1 | 10 | 1233 | |||
XM_005246947.2 | XP_005247004.1 | 10 | 1233 | |||
XM_005246948.2 | XP_005247005.1 | 10 | 1233 | |||
XM_006712838.3 | XP_006712901.1 | 10 | 1233 | |||
XM_011512148.2 | XP_011510450.1 | 10 | 1233 | |||
XM_017005252.1 | XP_016860741.1 | 10 | 1233 | |||
GenBank mRNA | AK097086.1 | - | 10 | 1052 | ||
AK301209.1 | - | 9 | 1164 | |||
AL133087.1 | - | 4 | 442 | |||
BC016985.2 | - | 10 | 1217 | 4395011 | ||
BC050586.2 | - | 7 | 869 | |||
BC063622.1 | - | 10 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
