siRNA Details
Gene Symbol: ZNF773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304334.1 | NP_001291263.1 | 4 | 1579 | ||
NM_001304336.1 | NP_001291265.1 | 4 | 1575 | |||
NM_198542.2 | NP_940944.1 | 4 | 1582 | |||
GenBank mRNA | AK057209.1 | - | 4 | 1498 | ||
BC005167.2 | - | 4 | 1495 | 3535599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
