siRNA Details
Gene Symbol: PDZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178140.3 | NP_835260.2 | 25 | 10804 | ||
XM_005248269.4 | XP_005248326.1 | 26 | 10928 | |||
XM_005248271.1 | XP_005248328.1 | 25 | 10280 | |||
XM_005248272.3 | XP_005248329.1 | 25 | 10282 | |||
XM_006714460.2 | XP_006714523.1 | 25 | 10079 | |||
XM_011513992.2 | XP_011512294.1 | 27 | 11290 | |||
XM_011513993.2 | XP_011512295.1 | 26 | 11208 | |||
XM_011513994.2 | XP_011512296.1 | 27 | 11255 | |||
XM_011513995.2 | XP_011512297.1 | 26 | 11187 | |||
XM_011513996.2 | XP_011512298.1 | 25 | 11014 | |||
XM_017009245.1 | XP_016864734.1 | 23 | 10004 | |||
XM_017009246.1 | XP_016864735.1 | 27 | 11206 | |||
GenBank mRNA | AB002298.2 | - | 25 | 10804 | ||
AK128686.1 | - | 3 | 3990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
