siRNA Details
Gene Symbol: PDZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178140.3 | NP_835260.2 | 12 | 2398 | ||
XM_005248269.4 | XP_005248326.1 | 12 | 2420 | |||
XM_005248271.1 | XP_005248328.1 | 11 | 1772 | |||
XM_005248272.3 | XP_005248329.1 | 11 | 1774 | |||
XM_006714460.2 | XP_006714523.1 | 11 | 1571 | |||
XM_011513992.2 | XP_011512294.1 | 13 | 2782 | |||
XM_011513993.2 | XP_011512295.1 | 12 | 2700 | |||
XM_011513994.2 | XP_011512296.1 | 13 | 2783 | |||
XM_011513995.2 | XP_011512297.1 | 13 | 2781 | |||
XM_011513996.2 | XP_011512298.1 | 11 | 2506 | |||
XM_017009245.1 | XP_016864734.1 | 9 | 1496 | |||
XM_017009246.1 | XP_016864735.1 | 13 | 2698 | |||
GenBank mRNA | AB002298.2 | - | 12 | 2398 | ||
AF338650.1 | - | 7 | 1470 | |||
AK021922.1 | - | 1 | 2313 | |||
AK304804.1 | - | 11 | 1767 | |||
BX648242.1 | - | 7 | 8615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
