siRNA Details
Gene Symbol: PDZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178140.3 | NP_835260.2 | 25 | 9655 | ||
XM_005248269.4 | XP_005248326.1 | 26 | 9779 | |||
XM_005248271.1 | XP_005248328.1 | 25 | 9131 | |||
XM_005248272.3 | XP_005248329.1 | 25 | 9133 | |||
XM_006714460.2 | XP_006714523.1 | 25 | 8930 | |||
XM_011513992.2 | XP_011512294.1 | 27 | 10141 | |||
XM_011513993.2 | XP_011512295.1 | 26 | 10059 | |||
XM_011513994.2 | XP_011512296.1 | 27 | 10106 | |||
XM_011513995.2 | XP_011512297.1 | 26 | 10038 | |||
XM_011513996.2 | XP_011512298.1 | 25 | 9865 | |||
XM_017009245.1 | XP_016864734.1 | 23 | 8855 | |||
XM_017009246.1 | XP_016864735.1 | 27 | 10057 | |||
GenBank mRNA | AB002298.2 | - | 25 | 9659 | ||
AK128686.1 | - | 3 | 2843 | |||
AK294741.1 | - | 3 | 1180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
