siRNA Details
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 3, 4 | 1327 | ||
GenBank mRNA | BC034021.1 | - | 5 | 480 | ||
M27879.1 | - | 2, 3 | 148 | |||
M55422.1 | - | 3, 4 | 1326 | |||
M61867.1 | - | 2, 3 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
