siRNA Details
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 4 | 1472 | ||
GenBank mRNA | BC034021.1 | - | 5 | 625 | ||
M27879.1 | - | 3 | 293 | |||
M55422.1 | - | 4 | 1471 | |||
M61867.1 | - | 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
