siRNA Details
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 4 | 2237 | ||
GenBank mRNA | BC034021.1 | - | 9 | 1390 | ||
M55422.1 | - | 4 | 2236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
